Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD31/PECAM-1 Antibody (GT0010), Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP313523
This item is not returnable.
View return policy
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CD31/PECAM-1 | |
| Monoclonal | |
| Unconjugated | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| The immunogen used to generate this antibody corresponds to human CD31/PECAM-1 | |
| 100 μg | |
| Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
| 5175 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| GT0010 | |
| PBS | |
| Armenian Hamster | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction