Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD31/PECAM-1 Antibody (MEC13.3), mFluor Violet 500 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB6001475MFV500
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Cell Surface)Specifications
CD31/PECAM-1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
MEC13.3 | |
Flow Cytometry, Flow (Cell Surface) | |
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
This CD31/PECAM-1 Antibody (MEC13.3) was developed against mouse endothelial cell line T-end. | |
0.1 mL | |
Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
5175 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction