Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD31/PECAM-1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171663MFV500
Description
CD31/PECAM-1 Polyclonal antibody specifically detects CD31/PECAM-1 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
CD31/PECAM-1 | |
Polyclonal | |
Flow Cytometry | |
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
This CD31/PECAM-1 Antibody was developed against a genomic peptide made to an internal region of human CD31/PECAM1 (within residues 100-300) [Swiss-Prot P16284]. | |
0.1 mL | |
Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
5175 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction