Learn More
Epredia™ Lab Vision™ CD31/PECAM-1 (Endothelial Cell Marker) Ab-1, Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: JC/70A
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat and pig. Others not tested.
Epitope: Not determined
Immunogen: Membrane preparation of a spleen from a patient with hairy cell leukemia
Molecular Weight: 100kDa (endothelial cells) and 130kDa (platelets)
Positive Control: Placenta, Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not suitable)
- Immunofluorescence
Specifications
Specifications
| Antigen | CD31 (Endothelial Cell Marker) Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | JC/70A |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Membrane preparation of a spleen from a patient with hairy cell leukemia |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | Oncogenesis |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.