Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD31/PECAM-1 (Endothelial Cell Marker) Ab-6, Mouse Monoclonal Antibody

Catalog No. MS1873S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1873S 1 mL Concentrated
MS1873S0 100 μL Concentrated
MS1873S1 500 μL Concentrated
MS1873R1 1 mL Prediluted
MS1873R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1873S Supplier Epredia™ Supplier No. MS1873S

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD31/PECAM-1 (Endothelial Cell Marker) Ab-6, Mouse Monoclonal Antibody.

CD31 is a glycoprotein expressed on endothelial cells and in platelets. It is known to be involved in cell signaling and cell adhesion.

Host Species: Mouse

Clone: 1A10

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Extracellular domain downstream of the signal sequence

Immunogen: Recombinant protein corresponding to the extracellular domain downstream of the signal sequence of the CD31 molecule

Molecular Weight: 100kDa (endothelial cells) and 130kDa (platelets)

Positive Control: Tonsil, Placenta, Liver, Lung

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD31 (Endothelial Cell Marker) Ab-6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A10
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to the extracellular domain downstream of the signal sequence of the CD31 molecule
Purification Method Tonsil
Quantity 1 mL
Regulatory Status RUO
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.