Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine CD31/PECAM-1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB33871R100UG
This item is not returnable.
View return policy
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Porcine samples. It is validated for Flow Cytometry.Specifications
CD31/PECAM-1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q95242 | |
PECAM1 | |
Mouse myeloma cell line NS0-derived recombinant porcine CD31/PECAM-1 Gln28-Lys602 (predicted) Accession # Q95242 | |
RUO | |
5175 | |
Pig | |
IgG1 |
Flow Cytometry | |
377537 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
Rat | |
100 μg | |
Primary | |
Detects porcine CD31/PECAM-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CD31, recombinant mouse (rm) CD31, rhVCAM-1, rhICAM-1, rhICAM-2, rhICAM-3, rmICAM-5, or rmMADCAM-1 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction