Learn More
Invitrogen™ CD31 (PECAM-1) Recombinant Mouse Monoclonal Antibody (BAG-85D10)

Description
Specificity: This antibody recognises CD31 (PECAM1) which is a member of the immunoglobulin superfamily and is expressed at various circulatng cells, such as neutrophils, monocytes, platelets as well as on endothelial cells. CD31 is a cell adhesion molecule and is involved in the leukocyte transendothelial migration during inflammation.
Specifications
Specifications
| Antigen | CD31 (PECAM-1) |
| Applications | Immunohistochemistry |
| Classification | Recombinant Monoclonal |
| Clone | BAG-85D10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | Pecam1 |
| Gene Accession No. | P16284 |
| Gene Alias | C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.