Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Rat anti-Mouse, FITC, Clone: MEC 13.3, BD

Catalog No. BDB553372 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553372 0.5 mg
BDB561813 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB553372 Supplier BD Biosciences Supplier No. 553372
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MEC13.3 antibody reacts with CD31, also known as PECAM-1 (Platelet Endothelial Cell Adhesion Molecule-1). CD31 is a 130 kDa integral membrane protein, a member of the immunoglobulin superfamily, that mediates cell-to-cell adhesion. CD31 is expressed constitutively on the surface of adult and embryonic endothelial cells and is also expressed on many peripheral leukocytes and platelets. It has also been detected on bone marrow-derived hematopoietic stem cells and embryonic stem cells. CD31 is involved in the transendothelial emigration of neutrophils, and neutrophil PECAM-1 appears to be down-regulated after extravasation into inflamed tissues. Multiple alternatively spliced isoforms are detected during early post-implantation embryonic development; this alternative splicing is involved in the regulation of ligand specificity. CD38 and vitronectin receptor (αvβ3 integrin, CD51/CD61) are proposed to be ligands for CD31. CD31-mediated endothelial cell-cell interactions are involved in angiogenesis. The MEC13.3 mAb inhibits a variety of in vitro and in vivo functions mediated by CD31.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31
Applications Flow Cytometry
Classification Monoclonal
Clone MEC 13.3
Concentration 0.5mg/mL
Conjugate FITC
Description PECAM-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen 129/Sv mouse-derived endothelioma cell line tEnd.1
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.