Learn More
CD31 Rat anti-Mouse, PE, Clone: MEC 13.3, BD
Rat Monoclonal Antibody
$68.00 - $344.00
Specifications
Antigen | CD31 |
---|---|
Clone | MEC 13.3 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The MEC13.3 antibody reacts with CD31, also known as PECAM-1 (Platelet Endothelial Cell Adhesion Molecule-1). CD31 is a 130 kDa integral membrane protein, a member of the immunoglobulin superfamily, that mediates cell-to-cell adhesion. CD31 is expressed constitutively on the surface of adult and embryonic endothelial cells and is also expressed on many peripheral leukocytes and platelets. It has also been detected on bone marrow-derived hematopoietic stem cells and embryonic stem cells. CD31 is involved in the transendothelial emigration of neutrophils, and neutrophil PECAM-1 appears to be down-regulated after extravasation into inflamed tissues. Multiple alternatively spliced isoforms are detected during early post-implantation embryonic development; this alternative splicing is involved in the regulation of ligand specificity. CD38 and vitronectin receptor (αvβ3 integrin, CD51/CD61) are proposed to be ligands for CD31. CD31-mediated endothelial cell-cell interactions are involved in angiogenesis. The MEC13.3 mAb inhibits a variety of in vitro and in vivo functions mediated by CD31.
Specifications
CD31 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
129/Sv mouse-derived endothelioma cell line tEnd.1 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
MEC 13.3 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
PECAM-1 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.