Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD31 Rat anti-Mouse, RB780, Clone: MEC 13.3, BD Horizon™
Rat Monoclonal Antibody
$98.40 - $253.50
Specifications
| Antigen | CD31 |
|---|---|
| Clone | MEC 13.3 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD31 Monoclonal antibody specifically detects CD31 in Mouse samples. It is validated for Flow cytometrySpecifications
| CD31 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 18613 | |
| IgG2a κ | |
| Affinity Chromatography |
| MEC 13.3 | |
| Flow Cytometry | |
| RB780 | |
| Rat | |
| Mouse | |
| EndoCAM; GPIIA; PECA1; PECAM1; Platelet endothelial cell adhesion molecule | |
| 129/Sv mouse-derived endothelioma cell line tEnd.1 | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title