Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD314 (NKG2D) Monoclonal Antibody (C7), Functional Grade, eBioscience™

Armenian Hamster Monoclonal Antibody

$258.00

Specifications

Antigen CD314 (NKG2D)
Clone C7
Concentration 1 mg/mL
Content And Storage 4°C
Applications Flow Cytometry, Functional Assay, Neutralization
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-145-84
Encompass_Preferred
View Documents
Invitrogen™
16587382
100 μg
Each of 1 for $258.00
Only null left
Add to Cart
 
Description

Description

Description: The C7 monoclonal antibody reacts with the mouse NKG2D. NKG2D is a lectin-like molecule expressed on both human and mouse NK cell lineage. mouse NKG2D binds to retinoic acid-inducible RAE1-alpha, -beta, -gamma, -delta, and -ε and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1^+ thymocytes, in vitro activated LAK cells and a subset of splenic NKT cells. A10 and C7 antibodies detect NK cells from all inbred strains of mouse tested so far. C7 is reported to interfere with the interaction of NKG2D with its ligands as shown by inhibition of lysis of Ba/F3-RAE1d by C57BL/6 LAK cells in the presence of the C7 mAb. C7 and another hamster anti-mouse NKG2D (clone A10, Product # 14-5872) compete with each other for binding to transfected cells by flow cytometric analysis, suggesting that they may bind to similar epitopes or block each other by steric hindrance. C7 (neutralizing) and A10 (activating) also exhibit different functional properties. Expression of the NKG2D antigen on mouse peripheral NK and NKT cells can be detected by flow cytometric analysis using mAb CX5 with much brighter intensity. Applications Reported: C7 has been reported for use in flow cytometric analysis, and in vitro blocking of mouse NKG2D (blocking). Applications Tested: This C7 antibody has been tested by flow cytometric ana...

Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. NK cells can regulate specific humoral and cell-mediated immunity, and preferentially express several calcium-dependent (C-type) lectins, which have been implicated in the regulation of NK cell function. The NK gene encodes a member of the NKG2 family, and the encoded transmembrane protein is characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The NKG2 gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed in NK cells.
Specifications

Specifications

CD314 (NKG2D)
1 mg/mL
Flow Cytometry, Functional Assay, Neutralization
Functional Grade
Armenian Hamster
RUO
PBS with no preservative; pH 7.2
O54709
27007
Primary
Affinity chromatography
C7
4°C
Monoclonal
Liquid
IgG
Mouse
Klrk1
CD314; D12S2489E; D6H12S2489E; killer cell lectin like receptor K1; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; KLR; Klrk1; natural killer cell group 2D; natural killer cell receptor; NK cell receptor D; NK lectin-like receptor; Nkg2d; NKG2-D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor; NKLLR; Nkrp2; NKR-P2; NKR-P2, ortholog of human NKG2D
Klrk1
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.