Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD314 (NKG2D) Monoclonal Antibody (CX5), Functional Grade, eBioscience™, Invitrogen™

Catalog No. 16588286
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-588-286 1 mg
50-145-98 100 μg
50-145-99 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16-588-286 Supplier Invitrogen™ Supplier No. 16588286
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The CX5 monoclonal antibody reacts with the mouse NKG2D, a lectin-like molecule expressed on both human and mouse NK cells. Mouse NKG2D binds to retinoic acid-inducible RAE-1alpha, -beta, -gamma, -delta, -ε and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors, through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1^+ thymocytes, in vitro activated LAK cells, and a subset of splenic NKT cells. Applications Reported: The CX5 antibody has been reported for use in flow cytometric analysis. It has also been reported in blocking of binding of NKG2D to its ligands, RAE-1 and H60, and inhibition NKG2D-dependent NK cell-mediated cytotoxicity against NKG2D ligand-bearing tumors in vitro and in vivo . Applications Tested: The CX5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that when using this antibody for staining the secondary must be Biotin anti-rat (cat 13-4813). Other secondaries may give suboptimal staining.

Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. NK cells can regulate specific humoral and cell-mediated immunity, and preferentially express several calcium-dependent (C-type) lectins, which have been implicated in the regulation of NK cell function. The NK gene encodes a member of the NKG2 family, and the encoded transmembrane protein is characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The NKG2 gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed in NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD314 (NKG2D)
Applications Flow Cytometry, Functional Assay, Neutralization
Classification Monoclonal
Clone CX5
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene Klrk1
Gene Accession No. O54709
Gene Alias CD314; D12S2489E; D6H12S2489E; killer cell lectin like receptor K1; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; KLR; Klrk1; natural killer cell group 2D; natural killer cell receptor; NK cell receptor D; NK lectin-like receptor; Nkg2d; NKG2-D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor; NKLLR; Nkrp2; NKR-P2; NKR-P2, ortholog of human NKG2D
Gene Symbols Klrk1
Host Species Rat
Purification Method Affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27007
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.