Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD314 (NKG2D) Monoclonal Antibody (1D11), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

$154.00 - $347.00

Specifications

Antigen CD314 (NKG2D)
Clone 1D11
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-109-30
Encompass_Preferred
View Documents
Invitrogen™
12587841
25 Tests
Each for $154.00
Only null left
Edge
Add to Cart
 
50-109-31
Encompass_Preferred
View Documents
Invitrogen™
12587842
100 Tests
Each for $347.00
Only null left
Edge
Add to Cart
 
Description

Description

Description: The 1D11 monoclonal antibody reacts with the human NKG2D, a 42 kDa lectin-like molecule expressed by NK cells, gamma delta T cells, CD8+ T cells, and some CD4+ T cells. Human NKG2D forms complexes with DAP10, a membrane adaptor protein, and has the ability to costimulate multiple NK activation receptors. The counter-receptor for human NKG2D has been identified as MICA/MICB expressed on epithelial tumors from lung, breast, kidney, ovary, prostate and colon carcinoma. 5C6 and 1D11 block binding of soluble MICA to gamma delta TCR T cell clones and inhibit lysis by these cells. 5C6 and 1D11 induced NKG2D function of redirected lysis of FcReceptor bearing P815 cells. Applications Reported: The 1D11 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1D11 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. NK cells can regulate specific humoral and cell-mediated immunity, and preferentially express several calcium-dependent (C-type) lectins, which have been implicated in the regulation of NK cell function. The NK gene encodes a member of the NKG2 family, and the encoded transmembrane protein is characterized by a type II membrane orientation (extracellular C terminus) and the presence of a C-type lectin domain. The NKG2 gene family is located within the NK complex, a region that contains several C-type lectin genes preferentially expressed in NK cells.
Specifications

Specifications

CD314 (NKG2D)
5 μL/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
CD314; D12S2489E; D6H12S2489E; killer cell lectin like receptor K1; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; KLR; Klrk1; natural killer cell group 2D; natural killer cell receptor; NK cell receptor D; NK lectin-like receptor; Nkg2d; NKG2-D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor; NKLLR; Nkrp2; NKR-P2; NKR-P2, ortholog of human NKG2D
Klrk1
Primary
4°C, store in dark, DO NOT FREEZE!
Klrk1
1D11
Flow Cytometry
PE
Mouse
Human
P26718
22914
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.