Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD317 (BST2) Mouse anti-Human, Rhesus, Baboon, Cynomolgus, Brilliant Violet 421, Clone: Y129 (also known as HM1.24), BD

Catalog No. BDB566382 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566382 25 μg
BDB566381 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566382 Supplier BD Biosciences Supplier No. 566382
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Y129 monoclonal antibody specifically binds to CD317 which is also known as Bone marrow stromal antigen 2 (BST2). CD317 is a type II transmembrane glycoprotein that belongs to the Tetherin family. CD317 is an interferon (IFN)-induced protein that is expressed on monocytes, granulocytes, dendritic cells, B lymphoblasts, plasma cells, T cells, natural killer (NK) cells, stromal cells and myeloma cells. CD317 is expressed by nonhematopoietic cells including cells within solid tumors derived from breast, lung and kidney. CD317 may be involved in the interactions between bone marrow stromal cells and lymphocytes. It is likewise known as Tetherin and reportedly blocks the release of some enveloped viruses by tethering virions to infected cell membranes. CD317 stimulates signaling by CD85g (ILT7) and may provide negative feedback for interferon (IFN) production by plasmacytoid dendritic cells during viral infections.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD317 (BST2)
Classification Monoclonal
Clone Y129 (also known as HM1.24)
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias BST2, BST-2, bone marrow stromal antigen 2, HM1.24 antigen, Tetherin
Host Species Mouse
Immunogen Human KPC-32 Plasma Cell Line
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.