Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD317 (BST2) Mouse anti-Human, Rhesus, Cynomolgus, Baboon, BUV395, Clone: Y129 (also known as HM1.24), BD Horizon™
Mouse Monoclonal Antibody
$182.00 - $442.00
Specifications
Antigen | CD317 (BST2) |
---|---|
Clone | Y129 (also known as HM1.24) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 395 |
Description
CD317 (BST2) Monoclonal antibody specifically detects CD317 (BST2) in Human, Rhesus, Cynomolgus, Baboon samples. It is validated for Flow cytometry (Routinely Tested)Specifications
CD317 (BST2) | |
Flow Cytometry | |
Brilliant Ultraviolet 395 | |
Mouse | |
Human, Rhesus Macaque, Cynomolgus, Baboon | |
BST2; BST-2; bone marrow stromal antigen 2; HM1.24 antigen; Tetherin | |
IgG2a κ | |
Affinity Chromatography |
Y129 (also known as HM1.24) | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human KPC-32 Plasma Cell Line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title