Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD317 (BST2) Mouse, anti-Mouse, R718, Clone: 927, BD OptiBuild™

Catalog No. BDB752140 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752140 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752140 Supplier BD Biosciences Supplier No. 752140
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 927 monoclonal antibody specifically recognizes CD317 which is also known as Bone marrow stromal antigen 2 (BST2) or Plasmacytoid Dendritic Cell Ag-1 (PDCA-1). CD317 (BST2) is a type II transmembrane glycoprotein that is encoded by Bst2. It is highly expressed on naïve plasmacytoid dendritic cells (pDC) which comprise very small populations within lymphoid and nonlymphoid tissues. pDC are characteristically early responders to viruses through Toll-like receptors (TLR), TLR7 and TLR9. Activated pDC can produce large amounts of proinflammatory cytokines including type I interferons, Interferon-alpha (IFN-α) and Interferon-beta (IFN-β), and are also known as Type I IFN producing cells (IPC). CD317 (BST2) expression can be upregulated on a variety of other cell types following stimulation with type I IFNs and IFN gamma including T cells, B cells, plasma cells, NK cells, CD8+ and CD8- dendritic cells, and cell lines. CD317 (BST2) may play a role in sorting proteins, eg, cytokines, from the Golgi apparatus and the plasma membrane.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD317 (BST2)
Applications Flow Cytometry
Classification Monoclonal
Clone 927
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.