Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD317 (BST2) Rat anti-Mouse, BUV661, Clone: 927, BD Biosciences™
Rat Monoclonal Antibody
Supplier: BD Biosciences 750635
Description
CD317 (BST2) Monoclonal antibody specifically detects CD317 (BST2) in Mouse samples. It is validated for Flow Cytometry
Specifications
CD317 (BST2) | |
Monoclonal | |
Brilliant Ultraviolet 661 | |
Bst2; BST-2; PDCA-1; PDCA1; Tetherin; DAMP-1; GREG | |
Mouse bone marrow-derived type I IFN-producing cella (IPC) | |
RUO | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
927 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction