Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD318 (CD6L), Mouse anti-Human, Clone: 3A11, Millipore Sigma™

Catalog No. MABF211925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF211925 25 μg
MABF2119100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF211925 Supplier MilliporeSigma Supplier No. MABF211925UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD318 (CD6L), clone 3A11, Cat. No. MABF2119, is a mouse monoclonal antibody that detects CD318 (CD6L) and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

CUB domain-containing protein 1 (UniProt: Q9H5V8; also known as Membrane glycoprotein gp140, Subtractive immunization M plus HEp3-associated 135 kDa protein, SIMA135, Transmembrane and associated with Src kinases, CD318, CD6L) is encoded by the CDCP1 (also known as TRASK) gene (Gene ID:64866) in human. CD318 is a single-pass membrane protein (isoform 1) that serves as a ligand for CD6. It is highly expressed in mitotic cells and has low expression during interphase. It is detected in high levels in skeletal muscle and colon tissue and in lower levels in kidney, placenta, lungs, and small intestine. Its levels are shown to be up-regulated in several human cancer tissues. It is also expressed in high levels in synovial tissues where it participates in CD6-dependent adhesion of T cells to synovial fibroblasts. CD318 is shown to be chemoattractive to T cells and levels of soluble CD318 are increased in the synovial fluid from patients with rheumatoid arthritis and juvenile inflammatory arthritis. CD318 is synthesized with a signal peptide (aa 1-29), which is cleaved off in the mature form. It has an extracellular domain (aa 30-667), a transmembrane domain (aa 668-688), and a cytoplasmic domain (aa 689-836). Three isoforms of CD318 have been described that are produced by alternative splicing. (Ref.: Enyindah-Asonye, G., et al. (2017). Proc. Natl. Acad. Sci. USA 114(33); E6912-E6921).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD318 (CD6L)
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3A11
Conjugate Unconjugated
Dilution Immunoprecipitation Analysis: A representative lot immunoprecipitated CD318 (CD6L) in Immunoprecipitation applications (Saifullah, M.K., et. al. (2004). J Immunol. 173(10):6125-33; Enyindah-Asonye, G., et. al. (2017). Proc Natl Acad Sci U S A. 114(33):E6912-E6921).Immunohistochemistry (Paraffin) Analysis: A representative lot detected CD318 (CD6L) in Immunohistochemistry applications (Enyindah-Asonye, G., et. al. (2017). Proc Natl Acad Sci U S A. 114(33):E6912-E6921).Western Blotting Analysis: A representative lot detected CD318 (CD6L) in Western Blotting applications (Saifullah, M.K., et. al. (2004). J Immunol. 173(10):6125-33; Enyindah-Asonye, G., et. al. (2017). Proc Natl Acad Sci U S A. 114(33):E6912-E6921).Flow Cytometry Analysis: A representative lot detected CD318 (CD6L) in Flow Cytometry applications (Saifullah, M.K., et. al. (2004). J Immunol. 173(10):6125-33; Enyindah-Asonye, G., et. al. (2017). Proc Natl Acad Sci U S A. 114(33):E6912-E6921).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias CUB domain-containing protein 1;Membrane glycoprotein gp140;Subtractive immunization M plus HEp3-associated 135 kDa protein;SIMA135;Transmembrane and associated with src kinases;CD6L
Gene Symbols CDCP1;TRASK
Host Species Mouse
Immunogen HBL-100 human mammary gland epithelial cells.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_073753.3
Test Specificity Clone 3A11 specifically detects CD318 in human cells. It targets an epitope with in the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.