Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD318 (CDCP1) Monoclonal Antibody (CSTEM26), eBioscience™, Invitrogen™

Catalog No. 14318937
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-318-937 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-318-937 Supplier Invitrogen™ Supplier No. 14318937
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This monoclonal antibody CSTEM26 reacts with human CD318, also known as CUB domain-containing protein 1 (CDCP1) and Trask. Applications Reported: This CSTEM26 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CSTEM26 antibody has been tested by flow cytometric analysis of normal human induced pluripotent stem cells. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

CDCP1 is a transmembrane protein containing three extracellular CUB domains. It is found to be overexpressed in colon and lung cancers. Its expression level is correlated with the metastatic ability of carcinoma cells. CDCP1 is located on the cell surface. It has been shown to be tyrosine phosphorylated in a cancer cell line. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD318 (CDCP1)
Applications Flow Cytometry
Classification Monoclonal
Clone CSTEM26
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cdcp1
Gene Accession No. Q9H5V8
Gene Alias 9030022E12Rik; AA409659; CD318; CDCP1; CUB domain containing protein 1; CUB domain-containing protein 1; E030027H19Rik; gp140; Membrane glycoprotein gp140; RGD1305578; SIMA135; Subtractive immunization M plus HEp3-associated 135 kDa protein; transmembrane and associated with src kinases; TRASK; UNQ2486/PRO5773
Gene Symbols Cdcp1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64866
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.