Learn More
Invitrogen™ CD319 (CRACC) Monoclonal Antibody (162), PE, eBioscience™

Description
Description: The monoclonal antibody 162 recognizes CRACC (CD2 like receptor for activating cytotoxic cells) also known as CD319 and CS1. It is a member of the SLAM receptor family. CRACC is a 66kDa glycosylated transmembrane protein which contains a cytoplasmic ITSM domain (Immunoreceptor Tyrosine-based Switch Motif). It is this domain, found in other proteins such as CD244 and NTB-A, that interacts with SH2 containing proteins such as SAP. Interestingly the ITSM domain in CRACC is SAP independent. Instead EAT-2 acts as the signaling molecule for CRACC. This has profound impact for patients with X-linked lymphoproliferative disease (contains a SAP mutation). It should be noted that a splice variant has been reported which deletes the ITSM domain. Applications Reported: This 162 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 162 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD319 (CRACC) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 162 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | SLAMF7 |
| Gene Accession No. | Q9NQ25 |
| Gene Alias | 19A; 19A24; 19A24 protein; 4930560D03Rik; CD2 subset 1; CD2-like receptor activating cytotoxic cells; CD2-like receptor-activating cytotoxic cells; CD319; CRACC; CS1; Leukocyte cell-surface antigen; membrane protein FOAP-12; Novel Ly9; novel LY9 (lymphocyte antigen 9) like protein; protein 19A; RGD1560190; SLAM family member 7; SLAMF7; UNQ576/PRO1138 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.