Learn More
CD319 (CRACC) Mouse anti-Human , PE, Clone: 162/CRACC (also known as 162), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD319 (CRACC) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 162/CRACC (also known as 162) |
| Conjugate | PE |
| Formulation | Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. |
| Gene Alias | 19A; 19A24 protein; CD319; CRACC; CS1; SLAMF7 |
| Host Species | Mouse |
| Immunogen | Human extracellular CRACC |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.