Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD32 Monoclonal Antibody (AT10)
GREENER_CHOICE

Catalog No. PIMA181209
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA181209 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA181209 Supplier Invitrogen™ Supplier No. MA181209
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

The epitope recognized by this antibody is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. A suggested positive control for immunohistochemical applications is lymph node. This product contains sodium azide; removal by dialysis is recommended prior to use in functional assays. Dialysis cassettes are suitable for this purpose.

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD32
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone AT10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.4
Gene FCGR2A
Gene Accession No. P12318, P31994, P31995
Gene Alias CD32; CD32 receptor 2; CD32 receptor 2a; CD32A; CD32B; CD32C; CDW32; Fc fragment of IgG receptor IIa; Fc fragment of IgG receptor IIb; Fc fragment of IgG receptor IIc (gene/pseudogene); Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIa, receptor (CD32); Fc fragment of IgG, low affinity IIa, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); Fc fragment of IgG, low affinity IIc, receptor for (CD32); Fc gamma receptor II; Fc gamma receptor IIa; Fc gamma receptor IIb; Fc gamma receptor IIC; Fc gamma RIIb; FCG2; Fc-gamma receptor II; fc-gamma RII; Fc-gamma RII-a; fc-gamma RII-b; Fc-gamma RII-c; fc-gamma-RIIa; Fc-gamma-RIIb; fc-gamma-RIIc; Fc-gamma-RII-D; FcGR; FCGR2; FCGR2A; FCGR2A1; FCGR2B; FCGR2C; fcRII; fcRII-a; fcRII-b; FCRIIC; FGFR2B; IGFR2; IgG Fc fragment receptor; IgG Fc gamma receptor II; igG Fc receptor II; IgG Fc receptor II-a; igG Fc receptor II-b; IgG Fc receptor II-c; Immunoglobulin G Fc receptor II; immunoglobulin G Fc receptor II-c; low affinity immunoglobulin gamma Fc region receptor II; Low affinity immunoglobulin gamma Fc region receptor II-a; low affinity immunoglobulin gamma Fc region receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II-c; MGC23887; MGC30032; RP11-5K23.6; soluble receptor; boFcRII-2
Gene Symbols FCGR2A, FCGR2B, FCGR2C
Host Species Mouse
Immunogen K562 cell line.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2212, 2213, 613131, 9103
Target Species Human, Canine, Monkey, Pig
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.