Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD32 Monoclonal Antibody (CCG39)
GREENER_CHOICE

Catalog No. PIMA528350
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528350 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528350 Supplier Invitrogen™ Supplier No. MA528350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Bovine CD32 antibody, clone CCG39, recognizes the bovine homologue of human CD32, one of a group of Fc receptors belonging to the immunoglobulin superfamily and involved in phagocytosis of opsonized microbes.

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD32
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone CCG39
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene FCGR2B
Gene Accession No. Q28110
Gene Alias CD32; CD32 receptor 2; CD32 receptor 2a; CD32A; CD32B; CD32C; CDW32; Fc fragment of IgG receptor IIa; Fc fragment of IgG receptor IIb; Fc fragment of IgG receptor IIc (gene/pseudogene); Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIa, receptor (CD32); Fc fragment of IgG, low affinity IIa, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); Fc fragment of IgG, low affinity IIc, receptor for (CD32); Fc gamma receptor II; Fc gamma receptor IIa; Fc gamma receptor IIb; Fc gamma receptor IIC; Fc gamma RIIb; FCG2; Fc-gamma receptor II; fc-gamma RII; Fc-gamma RII-a; fc-gamma RII-b; Fc-gamma RII-c; fc-gamma-RIIa; Fc-gamma-RIIb; fc-gamma-RIIc; Fc-gamma-RII-D; FcGR; FCGR2; FCGR2A; FCGR2A1; FCGR2B; FCGR2C; fcRII; fcRII-a; fcRII-b; FCRIIC; FGFR2B; IGFR2; IgG Fc fragment receptor; IgG Fc gamma receptor II; igG Fc receptor II; IgG Fc receptor II-a; igG Fc receptor II-b; IgG Fc receptor II-c; Immunoglobulin G Fc receptor II; immunoglobulin G Fc receptor II-c; low affinity immunoglobulin gamma Fc region receptor II; Low affinity immunoglobulin gamma Fc region receptor II-a; low affinity immunoglobulin gamma Fc region receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II-c; MGC23887; MGC30032; RP11-5K23.6; soluble receptor; boFcRII-2
Gene Symbols FCGR2A, FCGR2B
Host Species Mouse
Immunogen Bovine Fc gamma RII-transfected COS7 cells.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282229, 782652
Target Species Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.