Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD32 Mouse anti-Human, Brilliant Violet 421, Clone: 3D3, BD Optibuild™

Catalog No. BDB744253 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744253 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744253 Supplier BD Biosciences Supplier No. 744253
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 3D3 monoclonal antibody specifically recognizes FcγRII (CD32), a 40 kDa, polymorphic type I transmembrane glycoprotein that serves as a low affinity receptor for aggregated IgG. This highly glycosylated molecule (encoded by at least two different genes) is expressed on monocytes, granulocytes, platelets and B cells. Unlike the FLI28.26 mAb, the 3D3 mAb detected a polymorphic CD32 antigen expressed on B cells of all donors, but only on platelets, monocytes and granulocytes of some donors. The platelets from 3D3+ donors respond to certain stimulatory mAb such as CD165 (clone SN2) which results in aggregation. On the other hand, the platelets from 3D3 negative donors do not form aggregates after stimulation. Individuals can be divided into two groups as responder and non-responder depending on expression, or non-expression, of 3D3. In comparison to the 3D3 mAb, the FLI8.26 mAb detects a monomorphic CD32 antigen expressed on all human donors.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD32
Classification Monoclonal
Clone 3D3
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias FcγRII, CD32a/FcγRIIa/FCGR2A, CD32b/FcγRIIb/FCGR2B
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.