Learn More
CD32 Mouse anti-Human, BUV563, Clone: FLI8.26 (also known as 8.26), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD32 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | FLI8.26 (also known as 8.26) |
| Conjugate | Brilliant Ultraviolet 563 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | CD32a,FcγRIIa, FCGR2A; CD32b, FcγRIIb, FCGR2B; CD32c, FcγRIIc, FCGR2C |
| Host Species | Mouse |
| Immunogen | K562 or FcγRII+ L cell lines |
| Quantity | 50 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.