Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD32 Mouse anti-Rat, Unlabeled, Clone: D34-485, BD

Catalog No. BDB550270 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550270 0.1 mg
BDB550271 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB550270 Supplier BD Biosciences Supplier No. 550270
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The D34-485 antibody reacts specifically with CD32, the FcγII receptor. Rat CD32 is expressed on B lymphocytes, myeloid cells, and some lymphocytes in the thymic medulla. D34-485 mAb blocks binding of aggregated immunoglobulins to the FcγIi receptors in vitro. This antibody is routinely tested by flow cytometric analysis and in vitro blocking. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: D34-485
Isotype: IgG1 κ
Species Reactivity: Rat
Immunogen: Recombinant Rat CD32 Protein

Blocking Assay, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD32
Applications Flow Cytometry
Classification Monoclonal
Clone D34-485
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Fcgr2b; FcRII; Fc-gamma RII; FcγRII; FcγII receptor
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Rat CD32 Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.