Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587604
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587604 Supplier Invitrogen™ Supplier No. PA587604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TPAAPPKAVL KLEPQWINVL QEDSVTLTCR GTHSPESDSI PWFHNGNLIP THTQPSYRFK ANNNDSGEYT CQTGQTSLSD PVHLTVLSEW LVLQTPHLEF QEGETIVLRC HSWKDKPLVK VTFFQNGKSK KFSRSDPNFS IPQANHSHSG DYHCTGNIGY TLYSSKPVTI TVQAPSSSPM G; Positive Samples: THP-1, 22Rv1; Cellular Location: Cell membrane, Cytoplasm, Single-pass type I membrane protein.

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD32
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FCGR2A
Gene Accession No. P12318, P31994, P31995
Gene Alias CD32; CD32 receptor 2; CD32 receptor 2a; CD32A; CD32B; CD32C; CDW32; Fc fragment of IgG receptor IIa; Fc fragment of IgG receptor IIb; Fc fragment of IgG receptor IIc (gene/pseudogene); Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIa, receptor (CD32); Fc fragment of IgG, low affinity IIa, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); Fc fragment of IgG, low affinity IIc, receptor for (CD32); Fc gamma receptor II; Fc gamma receptor IIa; Fc gamma receptor IIb; Fc gamma receptor IIC; Fc gamma RIIb; FCG2; Fc-gamma receptor II; fc-gamma RII; Fc-gamma RII-a; fc-gamma RII-b; Fc-gamma RII-c; fc-gamma-RIIa; Fc-gamma-RIIb; fc-gamma-RIIc; Fc-gamma-RII-D; FcGR; FCGR2; FCGR2A; FCGR2A1; FCGR2B; FCGR2C; fcRII; fcRII-a; fcRII-b; FCRIIC; FGFR2B; IGFR2; IgG Fc fragment receptor; IgG Fc gamma receptor II; igG Fc receptor II; IgG Fc receptor II-a; igG Fc receptor II-b; IgG Fc receptor II-c; Immunoglobulin G Fc receptor II; immunoglobulin G Fc receptor II-c; low affinity immunoglobulin gamma Fc region receptor II; Low affinity immunoglobulin gamma Fc region receptor II-a; low affinity immunoglobulin gamma Fc region receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II-c; MGC23887; MGC30032; RP11-5K23.6; soluble receptor; boFcRII-2
Gene Symbols FCGR2A, FCGR2B, FCGR2C
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2212, 2213, 9103
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.