Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD320 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547155
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547155 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547155 Supplier Invitrogen™ Supplier No. PA547155
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Germinal center-B (GC-B) cells differentiate into memory B-cells and plasma cells (PC) through interaction with T-cells and follicular dendritic cells (FDC). CD320 augments the proliferation of PC precursors generated by IL-10. Receptor for the cellular uptake of transcobalamin bound cobalamin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD320
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Cd320
Gene Accession No. Q9NPF0
Gene Alias 425O18-1; 8D6; 8D6 antigen; 8D6A; 8D6AFDC-SM-8D6; 8D6TCBLR; Cd320; CD320 antigen; CD320 molecule; D17Ertd716e; FDC-signaling molecule 8D6; FDC-SM-8D6; NG29; putative VLDL lipoprotein receptor; RGD1305860; TCblR; transcobalamin receptor; UNQ198/PRO224; VLDL
Gene Symbols Cd320
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD320/TCblR/8D6A Leu31-Val231.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51293
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.