Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD320 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553182
Description
Immunogen sequence: AQERLRPLGL LVAMKESLLL SEQKTSLP Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.
Germinal center-B (GC-B) cells differentiate into memory B-cells and plasma cells (PC) through interaction with T-cells and follicular dendritic cells (FDC). CD320 augments the proliferation of PC precursors generated by IL-10. Receptor for the cellular uptake of transcobalamin bound cobalamin.
Specifications
CD320 | |
Polyclonal | |
Unconjugated | |
Cd320 | |
425O18-1; 8D6; 8D6 antigen; 8D6A; 8D6AFDC-SM-8D6; 8D6TCBLR; Cd320; CD320 antigen; CD320 molecule; D17Ertd716e; FDC-signaling molecule 8D6; FDC-SM-8D6; NG29; putative VLDL lipoprotein receptor; RGD1305860; TCblR; transcobalamin receptor; UNQ198/PRO224; VLDL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51293 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NPF0 | |
Cd320 | |
Recombinant protein corresponding to Human CD320. Recombinant protein control fragment (Product #RP-91028). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction