Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD320 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568014
Description
Predicted to react with Mouse and Rat samples.
Germinal center-B (GC-B) cells differentiate into memory B-cells and plasma cells (PC) through interaction with T-cells and follicular dendritic cells (FDC). CD320 augments the proliferation of PC precursors generated by IL-10. Receptor for the cellular uptake of transcobalamin bound cobalamin.
Specifications
CD320 | |
Polyclonal | |
Unconjugated | |
Cd320 | |
425O18-1; 8D6; 8D6 antigen; 8D6A; 8D6AFDC-SM-8D6; 8D6TCBLR; Cd320; CD320 antigen; CD320 molecule; D17Ertd716e; FDC-signaling molecule 8D6; FDC-SM-8D6; NG29; putative VLDL lipoprotein receptor; RGD1305860; TCblR; transcobalamin receptor; UNQ198/PRO224; VLDL | |
Rabbit | |
Affinity chromatography | |
RUO | |
362851, 51293, 54219 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5HZW5, Q9NPF0, Q9Z1P5 | |
Cd320 | |
Synthesized peptide derived from human CD320. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction