Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD324 (E-Cadherin) Mouse anti-Human, Brilliant Violet 786, Clone: 67A4, BD Optibuild™

Catalog No. BDB743716 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743716 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743716 Supplier BD Biosciences Supplier No. 743716
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 67A4 monoclonal antibody specifically recognizes the extracellular domain of human E-Cadherin (CD324). E-Cadherin is a 120-kDa transmembrane glycoprotein that is localized in the adherens junctions of epithelial cells. There it interacts with the cytoskeleton through the associated cytoplasmic catenin proteins. In addition to being a calcium-dependent adhesion molecule, E-Cadherin is also a critical regulator of epithelial junction formation. Its association with catenins is necessary for cell-to-cell adhesion. These E-Cadherin/catenin complexes associate with cortical actin bundles at both the zonula adherens and the lateral adhesion plaques. Tyrosine phosphorylation can disrupt these complexes, leading to changes in cell adhesion properties. E-Cadherin expression is often down-regulated in highly invasive, poorly differentiated carcinomas. Increased expression of E-Cadherin in these cells reduces their invasiveness. Thus, loss of expression or function of E-Cadherin appears to be an important step in tumorigenic progression. Pluripotent stem cells express E-Cadherin. Upon differentiation, an epithelial to mesenchymal transition results in the loss of E-cadherin expression and a gain in the expression of N-cadherin.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD324 (E-Cadherin)
Classification Monoclonal
Clone 67A4
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias E-cadherin, CD324, CDH1, CADH1, Cadherin-1, ECAD, CDHE, Arc-1, LCAM, UVO
Host Species Mouse
Immunogen Human Breast Tumor Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.