Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD324 (E-Cadherin) Rat anti-Mouse, Human, PE-CF594, Clone: DECMA-1, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 569555
Description
CD324 (E-Cadherin) Monoclonal antibody specifically detects CD324 (E-Cadherin) in Mouse|Human samples. It is validated for Flow cytometry
Specifications
CD324 (E-Cadherin) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
12550 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Flow Cytometry | |
DECMA-1 | |
PE-CF594 | |
ARC-1; Cdh1; E-cad; Ecad; UVO; cadherin-1; epithelial cadherin; uvomorulin | |
Mouse teratocarcinoma Cell Line | |
25 μg | |
Primary | |
Mouse, Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction