Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD325 (N-Cadherin) Recombinant Rabbit Monoclonal Antibody (BLR097G)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $523.50
Specifications
Antigen | CD325 (N-Cadherin) |
---|---|
Clone | BLR097G |
Concentration | 0.5 mg/mL |
Content And Storage | 4°C |
Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544385
|
Invitrogen™
MA544385 |
100 μL |
Each for $523.50
|
|
|||||
PIMA544386
|
Invitrogen™
MA544386 |
10 μL |
Each for $173.10
|
|
|||||
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
N-cadherin is a 140 kDa protein belonging to a family of transmembrane molecules that mediate calcium-dependent intercellular adhesion. Cadherins are involved in controlling morphogenetic movements during development and regulate cell surface adhesion through homotypic adhesion with the same cadherin species. N-cadherin's function is dependent on its association with the actin-cytoskeleton and is mediated through interactions between the C-terminal region of N-cadherin and the cytoplasmic catenin proteins. The stability of this association is regulated by phosphorylation and dephosphorylation of beta-catenin. Further, N-cadherin is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry.Specifications
CD325 (N-Cadherin) | |
0.5 mg/mL | |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
P19022 | |
1000 | |
Recombinant protein representing the extracellular domain of human N-Cadherin (Cadherin-2, residues 160-724). | |
Antibody |
BLR097G | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
CDH2 | |
bib; cadherin 2; cadherin 2 L homeolog; cadherin 2 type 1 N-cadherin (neuronal); cadherin 2, neuronal; cadherin 2, type 1 preproprotein; cadherin 2, type 1, N-cadherin (neuronal); cadherin 2, type 1, N-cadherin (neuronal) L homeolog; cadherin 2, type 1, N-cadherin a (neuronal); Cadherin2; cadherin-2; Cadherin-2A; Cadherin-N; calcium-dependent adhesion protein, neuronal; CD325; CDH2; cdh2.L; cdh2a; cdh2-a; cdh2-b; CDHN; cdhn-a; CDw325; CDw325 antigen; cell adhesion molecule; glass onion; glo; I79_014914; labyrinth; lyr; NCAD; N-Cad; N-cadherin; N-cadherin 1; N-cadherin A; N-cadherin precursor; N-cadherin-2; neural cadherin; Neural cadherin A; neural cadherin-2; neural calcium-dependent cell adhesion molecule; neural-cadherin; pac; Parachute; unnamed protein product; wu:fb47h04; XELAEV_18031953mg; ZNCAD | |
CDH2 | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title