Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD326 Rat anti-Mouse, BB515, Clone: G8.8, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 565425
Description
CD326 Monoclonal antibody specifically detects CD326 in Mouse samples. It is validated for Flow cytometry.
Specifications
CD326 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
G8.8 | |
Brilliant Blue 515 (BB515) | |
Ep-CAM; EGP; EGP-2; Egp314; GA733-2; TROP1; Tacsd1; Tacstd1; Ly74; gp40 | |
Glycoconjugates from BALB/c mouse-derived TE-71 medullary thymic epithelial cell line | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction