Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD326 Rat anti-Mouse, BUV805, Clone: G8.8, BD Biosciences™

Catalog No. BDB741996 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB741996 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB741996 Supplier BD Biosciences Supplier No. 741996
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD326 Monoclonal antibody specifically detects CD326 in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD326
Applications Flow Cytometry
Classification Monoclonal
Clone G8.8
Conjugate Brilliant Ultraviolet 805
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ep-CAM; EGP; EGP-2; Egp314; GA733-2; TROP1; Tacsd1; Tacstd1; Ly74; gp40
Host Species Rat
Immunogen Glycoconjugates from BALB/c mouse-derived TE-71 medullary thymic epithelial cell line
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimal conditions that minimize unconjugated dye and antibody.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.