Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD328 (Siglec7) Monoclonal Antibody (S7.7)
GREENER_CHOICE

Catalog No. PIMA528200
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528200 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528200 Supplier Invitrogen™ Supplier No. MA528200
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Human Siglec-7 antibody, cloneS7.7 recognizes human Sialic acid-binding Ig-like lectin 7, otherwise known as CD328, a putative adhesion molecule and member of the Ig superfamily, predominantly expressed by both resting and activated natural killer (NK) cells, and also at lower levels by monocytes, granulocytes, monocyte-derived macrophages and dendritic cells, and a small population of CD8+ memory T cells.

Putative adhesion molecule that mediates sialic-acid dependent binding to cells. Preferentially binds to alpha-2,3- and alpha-2,6-linked sialic acid. Also binds disialogangliosides (disialogalactosyl globoside, disialyl lactotetraosylceramide and disialyl GalNAc lactotetraoslylceramide). The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. In the immune response, may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. Mediates inhibition of natural killer cells cytotoxicity. May play a role in hemopoiesis. Inhibits differentiation of CD34+ cell precursors towards myelomonocytic cell lineage and proliferation of leukemic myeloid cells (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD328 (Siglec7)
Applications ELISA, Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S7.7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SIGLEC7
Gene Accession No. Q9Y286
Gene Alias Adhesion inhibitory receptor molecule 1; adhesion inhibitory receptor molecule 1, siglec-7; AIRM1; AIRM-1; CD328; CDw328; D-siglec; p75; p75/ AIRM-1; p75/AIRM1; QA79; QA79 membrane protein; sialic acid binding Ig like lectin 7; sialic acid binding Ig-like lectin 7; sialic acid binding immunoglobulin-like lectin 7; sialic acid-binding Ig-like lectin 7; Siglec; SIGLEC19P; SIGLEC7; SIGLEC-7; SIGLECP2
Gene Symbols SIGLEC7
Host Species Mouse
Immunogen Siglec-7 3T3 transfected cells.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27036
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.