Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD32b Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596718 Supplier Invitrogen™ Supplier No. PA596718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVALIYCRKK RISALPGYPE CREMGETLPE KPANPTNPDE ADKVGAENTI TYSLLMHPDA LEEPDDQNRI; Positive Samples: Human placenta; Cellular Location: Cell membrane, Single-pass type I membrane protein.

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD32b
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FCGR2B
Gene Accession No. P31994
Gene Alias AI528646; CD32; CD32 receptor 2; CD32 receptor 2a; CD32 receptor 2b; CD32A; CD32B; CDw32; F630109E10Rik; Fc fragment of IgG receptor IIb; Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor; Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); fc gamma receptor IIB; Fc gamma RIIb; Fc receptor, IgG, low affinity IIb; Fc[g]RII; FCG2; FC-gamma RII; fc-gamma RII-a; Fc-gamma RII-b; Fc-gamma-RII; fc-gamma-RIIa; FcgammaRIIB; fc-gamma-RIIb; FcgammaRIIB2; FcGR; Fcgr2; Fcgr2a; FCGR2A1; FCGR2B; FcgRII; Fcr-2; Fcr-3; fcRII; fcRII-a; fcRII-b; FGFR2B; IGFR2; IgG Fc receptor II beta; igG Fc receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; Ly-17; LyM-1; Ly-m20; Lymphocyte antigen 17
Gene Symbols FCGR2B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 241-310 of human FCGR2B (NP_0039923).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2213
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.