Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD32c (FCGR2C) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117462
Description
This antibody has been tested in direct-ELISA.
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
Specifications
CD32c (FCGR2C) | |
Polyclonal | |
Unconjugated | |
FCGR2C | |
AI528646; CD32; CD32C; CDW32; F630109E10Rik; Fc fragment of IgG receptor IIc (gene/pseudogene); Fc fragment of IgG, low affinity IIb, receptor; Fc fragment of IgG, low affinity IIc, receptor for (CD32); fc gamma receptor IIB; Fc gamma receptor IIC; Fc gamma RIIB; Fc receptor, IgG, low affinity IIb; Fc[g]RII; FCG2; fc-gamma RII; Fc-gamma RII-c; Fc-gamma-RII; FcgammaRIIB; fc-gamma-RIIB; FcgammaRIIB2; fc-gamma-RIIc; Fcgr2; Fcgr2a; Fcgr2b; FCGR2C; FcgRII; Fcr-2; Fcr-3; FcRII; FCRIIC; IGFR2; IgG Fc receptor II beta; IgG Fc receptor II-c; immunoglobulin G Fc receptor II-c; low affinity immunoglobulin gamma FC region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; low affinity immunoglobulin gamma Fc region receptor II-c; Ly-17; Lym-1; Ly-m20; Lymphocyte antigen 17 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9103 | |
-20°C or -80°C if preferred | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P31995 | |
FCGR2C | |
Recombinant Human Low affinity immunoglobulin gamma Fc region receptor II-c protein (43-223aa). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction