Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD32c (FCGR2C) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586699 Supplier Invitrogen™ Supplier No. PA586699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD32c (FCGR2C)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene FCGR2C
Gene Accession No. P31995
Gene Alias AI528646; CD32; CD32C; CDW32; F630109E10Rik; Fc fragment of IgG receptor IIc (gene/pseudogene); Fc fragment of IgG, low affinity IIb, receptor; Fc fragment of IgG, low affinity IIc, receptor for (CD32); fc gamma receptor IIB; Fc gamma receptor IIC; Fc gamma RIIB; Fc receptor, IgG, low affinity IIb; Fc[g]RII; FCG2; fc-gamma RII; Fc-gamma RII-c; Fc-gamma-RII; FcgammaRIIB; fc-gamma-RIIB; FcgammaRIIB2; fc-gamma-RIIc; Fcgr2; Fcgr2a; Fcgr2b; FCGR2C; FcgRII; Fcr-2; Fcr-3; FcRII; FCRIIC; IGFR2; IgG Fc receptor II beta; IgG Fc receptor II-c; immunoglobulin G Fc receptor II-c; low affinity immunoglobulin gamma FC region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; low affinity immunoglobulin gamma Fc region receptor II-c; Ly-17; Lym-1; Ly-m20; Lymphocyte antigen 17
Gene Symbols FCGR2C
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 278-323 of Human CD32-C.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9103
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.