Learn More
CD33 Monoclonal Antibody (FOS), Exalpha

Mouse Monoclonal Antibody
Supplier: Vector Laboratories 0331100UG


Description
Flow Cytometry PBMC: Add 10 μL of MAB/10^6 PBMC in 100 μL PBS. Mix gently and incubate for 15 minutes at 2 to 8C. Wash twice with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. Flow Cytometry Whole Blood: Add10 μL of MAB/100 μL of whole blood. Mix gently and incubate for 15 minutes at room temperature 20C. Lyse the whole blood. Wash once with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. See instrument manufacturer instructions for Lysed Whole Blood and Immunofluorescence analysis with a flow cytometer or microscope.
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow CytometrySpecifications
CD33 | |
Monoclonal | |
Unconjugated | |
CD33 | |
CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 | |
CD33 was derived from the hybridization of mouse Sp2/0 myeloma cells with spleen cells from BALB/c mice immunized with FMY9S5 cells containing the CD33 gene. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
FOS | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.2 | |
P20138 | |
Mouse | |
Protein A/G | |
RUO | |
945 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.