Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD33 Monoclonal Antibody (WM-53 (WM53)), NovaFluor™ Yellow 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$201.00 - $430.00
Specifications
Antigen | CD33 |
---|---|
Clone | WM-53 (WM53) |
Concentration | 0.8 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H015T03Y08A
![]() |
Invitrogen™
H015T03Y08A |
100 Tests |
Each for $430.00
|
|
|||||
H015T02Y08A
![]() |
Invitrogen™
H015T02Y08A |
25 Tests |
Each for $201.00
|
|
|||||
Description
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow CytometrySpecifications
CD33 | |
0.8 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P20138 | |
945 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
WM-53 (WM53) | |
Flow Cytometry | |
NovaFluor Yellow 755 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 | |
IgG1 κ | |
Affinity chromatography | |
CD33 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title