Learn More
Invitrogen™ CD33 Monoclonal Antibody (WM53), PE-Cyanine5.5

Description
CD33 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD33 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | WM53 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE-Cyanine5.5 |
| Formulation | PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.1 |
| Gene | CD33 |
| Gene Accession No. | P20138 |
| Gene Alias | CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.