Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD33 Mouse anti-Human, FITC, Clone: HIM3-4, BD

Catalog No. BDB561818 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561818 25 Tests
BDB555626 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561818 Supplier BD Biosciences Supplier No. 561818
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 67 kDa type I transmembrane glycoprotein expressed on monocytes, activated T cells, myeloid progenitors as well as mast cells. CD33 is absent on normal platelets, lymphocytes, erythrocytes and hematopoietic stem cells. Reports indicate that this glycoprotein can function as a sialic acid-dependent cell adhesion molecule. This function can be modulated by endogenous sialoglycoconjugates when CD33 is expressed on the membrane. HIM3-4 reacts with human and monkey granulocytes and monocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD33
Applications Flow Cytometry
Classification Monoclonal
Clone HIM3-4
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.