Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD33 Mouse anti-Human, RB780, Clone: P67.6, BD Horizon™

Mouse Monoclonal Antibody

$172.00 - $359.50

Specifications

Antigen CD33
Clone P67.6
Applications Flow Cytometry
Classification Monoclonal
Conjugate RB780
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB569096
View Documents
BD Biosciences
569096
100 Tests
Each for $359.50
Only null left
Add to Cart
 
BDB569097
View Documents
BD Biosciences
569097
25 Tests
Each for $172.00
Only null left
Add to Cart
 
Description

Description

CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow cytometry
Specifications

Specifications

CD33
Flow Cytometry
RB780
Mouse
Human
Siglec-3; SIGLEC3; Sialic acid-binding Ig-like lectin 3; p67; gp67; My9
CD33-transfected FMY9S5 Cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
P67.6
Monoclonal
Liquid
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
945
IgG1 κ
Affinity Chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.