Learn More
Invitrogen™ CD335 (NKp46) Monoclonal Antibody (9E2), PerCP-eFluor™ 710, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 46335942

Description
Description: This 9E2 monoclonal antibody reacts with human CD335, which is also known as NKp46. Expressed on NK cells, CD335 is a member of the natural cytotoxicity receptor family that also includes NKp30 and NKp44. This type I transmembrane glycoprotein is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor and virus-infected (e.g., influenza) cells. CD335 also interacts with pancreatic beta cells during the development of type I diabetes. The 9E2 antibody has been reported to induce cytotoxicity. Applications Reported: This 9E2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 9E2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 μL cell sample + 100 μL IC Fixa...
CD335, also known as NKp46 or NCR1, is a member of the natural cytotoxicity receptor family, which includes NKp30 and NKp44. This type I transmembrane glycoprotein is expressed on NK cells and plays a crucial role in activating and expanding resting NK cells through interactions with dendritic cells. CD335 enhances the ability of NK cells to eliminate tumor and virus-infected cells, such as those infected by influenza. CD335 consists of two Ig-like domains and is related to leukocyte immunoglobulin-like receptors (LIR) and killer inhibitory receptors (KIR). It contributes to the increased efficiency of activated NK cells in mediating tumor cell lysis. Additionally, CD335 interacts with pancreatic beta cells during the development of type 1 diabetes, highlighting its involvement in both immune defense and autoimmune processes.
Specifications
CD335 (NKp46) | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
O76036 | |
NCR1 | |
Affinity chromatography | |
RUO | |
9437 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
9E2 | |
PerCP-eFluor 710 | |
NCR1 | |
1-Mar; activating receptor 1; Ar1; CD335; hNKp46; Kilr1; KILR-1; LY94; Ly-94; lymphocyte antigen 94; Lymphocyte antigen 94 homolog; lymphocyte antigen 94 homolog (activating NK-receptor; lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46); mAR-1; MAR1 (mouse activating receptor 1); mNKp46; natural cytotoxicity triggering receptor 1; Natural killer cell p46-related protein; natural killer cell receptor; Ncr1; NK cell-activating receptor; NK receptor KILR-1; NKACTR; Nkp46; NK-p46; NK-p46); rAR-1; rat-activating receptor 1 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.