Learn More
CD336 (NKp44) Mouse anti-Human, Brilliant Violet 510, Clone: p44-8, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 744300
Description
The p44-8 monoclonal antibody specifically binds to the natural killer (NK) cell receptor, NKp44, which is also known as CD336, Natural cytotoxicity triggering receptor 2 (NCR2), or Lymphocyte antigen 95 homolog (Ly95). NKp44 is a ñ44 kDa type I transmembrane protein that belongs to the natural cytotoxicity receptor (NCR) family within the immunoglobulin superfamily. NKp44 is expressed by activated NK cells. NKp44 serves as an activating receptor that can enhance NK cell mediated lysis of target cells including tumor cells and virus-infected cells. Killer activating receptor associated protein (KARAP), which is also known as DAP12, is an intracellular adaptor protein that can associate with the intracellular region of NKp44. DAP12 can then function to help transmit activating signals through its immunoreceptor tyrosine-based activation motif (ITAM).
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications
CD336 (NKp44) | |
p44-8 | |
Brilliant Violet 510 | |
CD336, NCR2, NCTR2, NKp44, NK-p44, LY95, dJ149M18.1 | |
Human NKp44 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.