Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD336 (NKp44) Mouse anti-Human, BUV615, Clone: p44-8, BD OptiBuild™

Catalog No. BDB752353 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752353 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752353 Supplier BD Biosciences Supplier No. 752353
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD336 (NKp44) Monoclonal antibody specifically detects CD336 (NKp44) in Human samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD336 (NKp44)
Applications Flow Cytometry
Classification Monoclonal
Clone p44-8
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Hu NKp44 (CD336) BUV615 P44-8 50 μg
Host Species Mouse
Immunogen Human NKp44
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD336 (NKp44)
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1 κ
Reactivity Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.