Learn More
CD337 (NKp30) Monoclonal Antibody (AF29-4D12), PE-Cyanine7, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$444.00
Specifications
| Antigen | CD337 (NKp30) |
|---|---|
| Clone | AF29-4D12 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-9269
![]()
|
Invitrogen™
25337942 |
100 Tests |
Each of 1 for $444.00
|
|
|||||
Description
Description: This AF29-4D12 monoclonal antibody reacts with human CD337, which is also known as NKp30. Expressed on NK cells, CD337 is the predomit member of the natural cytotoxicity receptor family that also includes NKp46 and NKp44. CD337 is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor, virus-infected, and immature dendritic cells. Several ligands for this receptor have been reported, including B7-H6. The AF29-4D12 antibody has been reported to crossreact to rhesus macaque. Applications Reported: This AF29-4D12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AF29-4D12 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
The protein encoded by this gene is a natural cytotoxicity receptor (NCR) that may aid NK cells in the lysis of tumor cells. The encoded protein interacts with CD3-zeta (CD247), a T-cell receptor. A single nucleotide polymorphism in the 5' untranslated region of this gene has been associated with mild malaria suceptibility. Three transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, May 2010].Specifications
| CD337 (NKp30) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| O14931, Q8MJ02 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| AF29-4D12 | |
| Flow Cytometry | |
| PE-Cyanine7 | |
| Mouse | |
| Human, Monkey, Rhesus Macaque | |
| 259197, 715574 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.