Learn More
Invitrogen™ CD337 (NKp30) Monoclonal Antibody (AF29-4D12), PE, eBioscience™, Invitrogen™
Description
Description: This AF29-4D12 monoclonal antibody reacts with human CD337, which is also known as NKp30. Expressed on NK cells, CD337 is the predominant member of the natural cytotoxicity receptor family that also includes NKp46 and NKp44. CD337 is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor, virus-infected, and immature dendritic cells. Several ligands for this receptor have been reported, including B7-H6. The AF29-4D12 antibody has been reported to crossreact to rhesus macaque. Applications Reported: This AF29-4D12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AF29-4D12 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD337 (NKp30) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | AF29-4D12 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Ncr3 |
| Gene Accession No. | O14931, Q8MJ02 |
| Gene Alias | 1C7; Activating natural killer receptor p30; activating NK-A1 receptor; CD337; LY117; lymphocyte antigen 117; MALS; Natural cytotoxicity triggering receptor 3; natural killer cell p30-related protein; natural killer cell receptor; Ncr3; NK receptor 1c7; Nkp30; NK-p30; NKp30S; NKp30v1; NKp30v1ED |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.