Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD339 (Jagged 1) Monoclonal Antibody (HMJ1-29), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5010763
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-107-63 100 μg
50-107-62 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-107-63 Supplier Invitrogen™ Supplier No. 12339182
Add to Cart
Edge
Add to Cart

Armenian Hamster Monoclonal Antibody

Description: This HMJ1-29 monoclonal antibody reacts with mouse Jagged-1, one of five type I transmembrane proteins that serves as a Notch receptor ligand. Upon binding the Notch receptor (e.g. Notch 1-4), Jagged-1 undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases an intracellular fragment whose biological function remains controversial. Jagged-1 is expressed by thymic lymphoid and stromal cells, as well as macrophages and CD8+CD11c+ dendritic cells in the spleen. The HMJ1-29 antibody is reported to crossreact to rat. Applications Reported: This HMJ1-29 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HMJ1-29 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD339 (Jagged 1)
Applications Flow Cytometry
Classification Monoclonal
Clone HMJ1-29
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene JAG1
Gene Accession No. Q63722, Q9QXX0
Gene Alias ABE2; AGS; AHD; AWS; CD339; Gsfabe2; HJ1; Htu; JAG1; jagged 1; jagged canonical Notch ligand 1; Jagged1; JAGL1; Ozz; protein jagged-1; Ser-1; Serrate-1
Gene Symbols JAG1
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16449, 29146
Target Species Mouse, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.